Learn More
Triphenylamine-d15, CDN
Click to view available options
Quantity:
0.25 g
0.5 g
Unit Size:
0.25g
0.5g
Specifications
Specifications
| Chemical Name or Material | Triphenylamine-D15 |
| CAS | 880462-20-4 |
| Molecular Formula | C18 D15 N |
| Quantity | 0.5 g |
| Synonym | Benzen-d5-amine, N,N-di(phenyl-d5)- (9 CI), N,N-Di(phenyl-d5)benzen-d5-amine, Triphenylamine-d15, Triphenylamine-D15, N,N-Diphenylaniline-d15 |
| InChI Formula | InChI=1 S/C18H15N/c1-4-10-16(11-5-1)19(17-12-6-2-7-13-17)18-14-8-3-9-15-18/h1-15 H/i1D,2 D,3 D,4 D,5 D,6 D,7 D,8 D,9 D,10 D,11 D,12 D,13 D,14 D,15 D |
| SMILES | [2 H]c1c([2 H])c([2 H])c(N(c2c([2 H])c([2 H])c([2 H])c([2 H])c2[2 H])c3c([2 H])c([2 H])c([2 H])c([2 H])c3[2 H])c([2 H])c1[2 H] |
| IUPAC Name | 2,3,4,5,6-pentadeuterio-N,N-bis(2,3,4,5,6-pentadeuteriophenyl)aniline |
| Molecular Weight (g/mol) | 260.411 |
| Formula Weight | 260.215 g/mol |
| Show More |