Learn More
Leica Microsystems Trinocular Tube
Product Code. 11974499
Change view
Click to view available options
For Use With (Equipment):
DM IL LED Inverted Microscope
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) unitSize
11974499 DM IL LED Inverted Microscope Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11974499 Supplier Leica Microsystems Supplier No. 11521249
Add to basket
Add to basket
A new inverted microscope for routine and laboratory microscopy in cell biology and medicine
The Leica DM IL LED is a new inverted microscope for routine and laboratory microscopy in cell biology and medicine.
  • The perfect choice for applications in cell biology and medicine, micromanipulation, biotechnology, developmental biology, transgenics, molecular biology and fluorescence
  • Sturdy design with low stage height and plenty of space for operation
  • Long working distance objectives
  • Ergonomic arrangement of controls for fatigue free operation
  • Outstanding phase contrast for unstained samples
  • LED illumination provides low energy consumption and no heat build up
  • LEDs last 50,000 hours, eliminating the inconvenience of replacing lamps
TRUSTED_SUSTAINABILITY
Specifications
Type Trinocular Tube
For Use With (Application) Tube Program DM
For Use With (Equipment) DM IL LED Inverted Microscope
Quantity 1 Ea.