Learn More
Motic™ Trinocular Eyepiece Adapter, 28 Series
Product Code. 15415112
Change view
Click to view available options
For Use With (Equipment):
Series 28 Microscopes
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
15415112 Series 28 Microscopes Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15415112 Supplier Motic™ Supplier No. 1101000300592
  • ONLINE EXCLUSIVE PRICE
    85.30€
    83.00€ / Pack of 1
    Save 2.30€
    2% Off
Estimated Shipment: 28-12-2026
Facilitate photomicography with this adapter. Motic™ Trinocular Eyepiece Adapter, 28 Series allows you to connect a C-type SLR camera to your microscope.
  • Facilitates photomicography.
  • Designed for C-mount SLR cameras.
Specifications
Type C-type and SLR Camera Adapter
For Use With (Equipment) Series 28 Microscopes
Includes Eyepiece