Learn More
Trimethylsulfonium Chloride, TRC
Product Code. 30453639
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30453639 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30453639 Supplier Toronto Research Chemicals Supplier No. TRCT7983751G
 Restricted Item
Estimated Shipment: 14-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Trimethylsulfonium Chloride
CAS 3086-29-1
Molecular Formula C3H9S . Cl
Quantity 1 g
Synonym Sulfonium','trimethyl-','chloride (1:1) (ACI), Sulfonium','trimethyl-','chloride (8CI','9CI), Trimethylsulfonium chloride (6CI','7CI)
InChI Formula InChI=1S/C3H9S.ClH/c1-4(2)3;/h1-3H3;1H/q+1;/p-1
SMILES [Cl-].C[S+](C)C
IUPAC Name trimethylsulfanium;chloride
Molecular Weight (g/mol) 112.62
Formula Weight 112.0113