Learn More
Trimethylolpropane Triglycidyl Ether (Technical Grade), TRC
Product Code. 30008956
Change view
Click to view available options
Quantity:
500 mg
1 g
5 g
Unit Size:
1g
500mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30008956 1 g 1g
30005939 5 g 5g
30003477 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30008956 Supplier Toronto Research Chemicals Supplier No. TRCT2193251G
 Restricted Item
Estimated Shipment: 12-11-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 30499-70-8
Molecular Formula C15H26O6
Molecular Weight (g/mol) 302.36
Synonym 2-Ethyl-2-(hydroxymethyl)-1,3-propanediol Polymer (Chloromethyl)oxirane;Denacol EX 320, Epichlorohydrin-trimethylolpropane Copolymer, Epichlorohydrin-trimethylolpropane Polymer, Oksilin 5, T 856506, Triepoxide 427-61;Trimethylolpropane-epichlorohydrin Copolymer;
IUPAC Name 2-[2,2-bis(oxiran-2-ylmethoxymethyl)butoxymethyl]oxirane
SMILES CCC(COCC1CO1)(COCC2CO2)COCC3CO3
Specifications
Quantity 1 g
InChI Formula InChI=1S/C15H26O6/c1-2-15(9-16-3-12-6-19-12,10-17-4-13-7-20-13)11-18-5-14-8-21-14/h12-14H,2-11H2,1H3
Formula Weight 302.1729
Chemical Name or Material Trimethylolpropane Triglycidyl Ether (Technical Grade)