missing translation for 'onlineSavingsMsg'
Learn More

Trimethyl thiophosphate

Product Code. 11377268
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 11377268

Brand: Thermo Scientific Alfa Aesar 030395.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 152-18-1 | C3H9O3PS | 156.136 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 152-18-1
Molecular Formula C3H9O3PS
Molecular Weight (g/mol) 156.136
MDL Number MFCD00014888
InChI Key XWSLYQXUTWUIKM-UHFFFAOYSA-N
Synonym trimethyl thiophosphate, trimethylthiophosphate, o,o,o-trimethyl phosphorothioate, o,o,o-trimethyl thiophosphate, trimethylthiofosfat, o,o,o-trimethylthiofosfat, trimethylthiofosfat czech, ccris 5569, phosphorothioic acid, o,o,o-trimethyl ester, o,o,o-trimethylthiofosfat czech
PubChem CID 9038
IUPAC Name trimethoxy(sulfanylidene)-$l^{5}-phosphane
SMILES COP(=S)(OC)OC

Specifications

Color White
Odor Sulphurous
Quantity 25 g
UN Number UN3278
Formula Weight 156.15
Physical Form Liquid
Chemical Name or Material Trimethyl thiophosphate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.