Learn More
Trimethyl-d9-sulfonium Iodide, CDN
Product Code. 30312333
Click to view available options
Quantity:
0.25 g
0.5 g
Unit Size:
0.25g
0.5g
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30312333
Brand: CDN CDND60930.25G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Trimethyl-d9-sulfonium Iodide
CAS 106776-17-4
Molecular Formula C3 D9 S . I
Quantity 0.25 g
Synonym Tri(methyl-d3)sulfonium Iodide, Sulfonium, tri(methyl-d3)-, iodide (9 CI)
InChI Formula InChI=1 S/C3H9S.HI/c1-4(2)3;/h1-3H3;1 H/q+1;/p-1/i1D3,2D3,3D3;
SMILES [I-].[2 H]C([2 H])([2 H])[S+](C([2 H])([2 H])[2 H])C([2 H])([2 H])[2 H]
IUPAC Name tris(trideuteriomethyl)sulfanium;iodide
Molecular Weight (g/mol) 213.1285
Formula Weight 213.0035 g/mol
Percent Purity 99 atom % D, min 98% Chemical Purity