Learn More
Mapa Professional Solo 980 Disposable Glove
Product Code. 30533548
Click to view available options
Size:
6
7
8
9
10
11
Unit Size:
25 pairs
Product Code. 30533548
Brand: Mapa Professional 34980458
Thick nitrile disposable gloves that offer both dexterity and resistance, suitable for various applications.
Comfort:
  • Designed for comfort and flexibility
Productivity:
  • Pebbled fingertips provide enhanced grip
Additional Benefits:
  • Delivers reliable protection against a broad range of chemicals, including isopropanol and certain Skydrol™ and Diestone™ formulations
Skin Protection:
  • Latex-free to minimize allergy risks
  • Powder-free to reduce contamination
  • Recommended for use in oily industrial environments
Standards:
  • EN 374 Type B: JKPT
  • EN 421:2010
  • Suitable for food contact (Alimentary - Europe)
  • Category III
  • Risk Types: Liquid, Chemical, Alimentary
Specifications
Certifications/Compliance (Europe) CE Category 3
Accelerator Free No
Applications Industrial, Laboratory, Healthcare
AQL 1.5
Color Blue
Disposable Single-use
Exterior Finish Fully Textured
Finger Thickness (English) 7.8 mil
Finger Thickness (Metric) 0.20 mm
Latex Free Yes
Material Nitrile
Palm Thickness (English) 7.8 mil
Palm Thickness (Metric) 0.20 mm
Powdered Powder Free
Product Type Nitrile Gloves
Size 8
Sterility Non-sterile
Thickness (English) 7.8 mil
Thickness (Metric) 0.20 mm
Glove Cuff Style Rolled
Resistant To Chemicals
Length (Metric) 30 cm
Product Line Solo 980