missing translation for 'onlineSavingsMsg'
Learn More

Trilaurin, 98%

Product Code. 10330344 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 10330344

Brand: Thermo Scientific Acros 421831000

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products

This item is not returnable. View return policy

CAS: 538-24-9 | C39H74O6 | 639.01 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 538-24-9
Molecular Formula C39H74O6
Molecular Weight (g/mol) 639.01
MDL Number MFCD00026559
InChI Key VMPHSYLJUKZBJJ-UHFFFAOYSA-N
Synonym trilaurin, glycerol trilaurate, glyceryl tridodecanoate, glyceryl trilaurate, glycerin trilaurate, lauric acid triglyceride, tridodecanoin, lauric acid triglycerin ester, trilauroylglycerol, laurin, tri
PubChem CID 10851
ChEBI CHEBI:77389
IUPAC Name 2,3-di(dodecanoyloxy)propyl dodecanoate
SMILES CCCCCCCCCCCC(=O)OCC(COC(=O)CCCCCCCCCCC)OC(=O)CCCCCCCCCCC

Specifications

Melting Point 45°C to 47°C
Color White
Density 0.9000g/mL
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Quantity 100 g
Specific Gravity 0.9
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in ethanol,diethyl ether,very soluble in acetone,benzene
Formula Weight 639.01
Percent Purity 98%
Physical Form Powder
Chemical Name or Material Trilaurin
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.