Learn More
Triisopropylsilanol, TRC
Product Code. 30420638
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30420638 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30420638 Supplier Toronto Research Chemicals Supplier No. TRCT2198681G
 Restricted Item
Estimated Shipment: 15-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Triisopropylsilanol
CAS 17877-23-5
Molecular Formula C9 H22 O Si
Quantity 1 g
Synonym 1,1,1-Tris(1-methylethyl)silanol
InChI Formula InChI=1S/C9H22OSi/c1-7(2)11(10,8(3)4)9(5)6/h7-10H,1-6H3
SMILES CC(C)[Si](O)(C(C)C)C(C)C
IUPAC Name hydroxy-tri(propan-2-yl)silane
Molecular Weight (g/mol) 174.356
Formula Weight 174.144