missing translation for 'onlineSavingsMsg'
Learn More

Triethylsulfonium iodide, 97%

Product Code. 11310164
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 11310164

Brand: Thermo Scientific Alfa Aesar L00667.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1829-92-1 | C6H15IS | 246.15 g/mol

Triethylsulfonium iodide is used as an ethylidene-transfer reagent. It is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Triethylsulfonium iodide is used as an ethylidene-transfer reagent. It is used as a pharmaceutical intermediate.

Solubility
It is soluble in water.

Notes
Light, Heat Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1829-92-1
Molecular Formula C6H15IS
Molecular Weight (g/mol) 246.15
MDL Number MFCD00051867
InChI Key LKDQWVKWYGOVJW-UHFFFAOYSA-M
Synonym triethylsulfonium iodide, triethylsulfanium iodide, triethylsulphonium iodide, sulfonium, triethyl-, iodide, acmc-20aje7, sulfonium,triethyl-,iodide
PubChem CID 74589
IUPAC Name triethylsulfanium;iodide
SMILES CC[S+](CC)CC.[I-]

Specifications

Melting Point ∽148°C (decomposition)
Quantity 25 g
Beilstein 3911790
Sensitivity Light sensitive
Solubility Information It is soluble in water.
Formula Weight 246.15
Percent Purity 97%
Chemical Name or Material Triethylsulfonium iodide

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.