Learn More
Triethylsilane-d1, TRC
Specifications
Specifications
| Chemical Name or Material | Triethylsilane-d1 |
| CAS | 1631-33-0 |
| Molecular Formula | C6H15DSi |
| Quantity | 250 mg |
| Synonym | Triethylsilane-d, Deuteriotriethylsilane, Deuterotriethylsilane, Triethyl-silane-d |
| InChI Formula | InChI=1S/C6H16Si/c1-4-7(5-2)6-3/h7H,4-6H2,1-3H3/i7D |
| SMILES | [2H][Si](CC)(CC)CC |
| IUPAC Name | deuterio(triethyl)silane |
| Molecular Weight (g/mol) | 117.28 |
| Formula Weight | 117.1084 |