Learn More
Triethylsilane-d1, TRC
Product Code. 30449830
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30449830 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30449830 Supplier Toronto Research Chemicals Supplier No. TRCT77740250MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Triethylsilane-d1
CAS 1631-33-0
Molecular Formula C6H15DSi
Quantity 50 mg
Synonym Triethylsilane-d, Deuteriotriethylsilane, Deuterotriethylsilane, Triethyl-silane-d
InChI Formula InChI=1S/C6H16Si/c1-4-7(5-2)6-3/h7H,4-6H2,1-3H3/i7D
SMILES [2H][Si](CC)(CC)CC
IUPAC Name deuterio(triethyl)silane
Molecular Weight (g/mol) 117.28
Formula Weight 117.1084