missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Triethyl-d15-amine, CDN
Click to view available options
Quantity:
0.25 g
0.5 g
Unit Size:
0.25g
0.5g
Specifications
Specifications
| Chemical Name or Material | Triethylamine-d15 |
| CAS | 66688-79-7 |
| Molecular Formula | C6 2H15 N |
| Quantity | 0.25 g |
| InChI Formula | InChI=1 S/C6H15N/c1-4-7(5-2)6-3/h4-6H2,1-3H3/i1D3,2D3,3D3,4D2,5D2,6D2 |
| SMILES | [2 H]C([2 H])([2 H])C([2 H])([2 H])N(C([2 H])([2 H])C([2 H])([2 H])[2 H])C([2 H])([2 H])C([2 H])([2 H])[2 H] |
| IUPAC Name | 1,1,2,2,2-pentadeuterio-N,N-bis(1,1,2,2,2-pentadeuterioethyl)ethanamine |
| Molecular Weight (g/mol) | 116.282 |
| Formula Weight | 116.215 g/mol |
| Percent Purity | 99 atom % D, min 98% Chemical Purity |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction