Learn More
Triclosan Methyl-d3 Ether, TRC
Specifications
Specifications
| Chemical Name or Material | TriclosanmethylD3(methoxyD3) |
| CAS | 1020720-00-6 |
| Molecular Formula | C13 2H3 H6 Cl3 O2 |
| Quantity | 50 mg |
| Synonym | Benzene','4-chloro-1-(2,4-dichlorophenoxy)-2-(methoxy-d3)-, Triclosan methyl D3 (methoxy D3), Triclosan Methyl-d3 Ether, Triclosan-methyl ether D3 (methoxy D3) |
| InChI Formula | InChI=1S/C13H9Cl3O2/c1-17-13-7-9(15)3-5-12(13)18-11-4-2-8(14)6-10(11)16/h2-7H,1H3/i1D3 |
| SMILES | [2H]C([2H])([2H])Oc1cc(Cl)ccc1Oc2ccc(Cl)cc2Cl |
| IUPAC Name | 2,4-dichloro-1-[4-chloro-2-(trideuteriomethoxy)phenoxy]benzene |
| Molecular Weight (g/mol) | 306.59 |
| Formula Weight | 304.9856 |