Learn More
Trichlorooctyl-stannane, TRC
Product Code. 30007640
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30007640 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30007640 Supplier Toronto Research Chemicals Supplier No. TRCT7743805G
 Restricted Item
Estimated Shipment: 17-11-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
CAS 3091-25-6
Molecular Formula C8 H17 Cl3 Sn
Quantity 5 g
Synonym Octyltin trichloride, Trichloro(n-octyl)stannane
InChI Formula InChI=1S/C8H17.3ClH.Sn/c1-3-5-7-8-6-4-2;;;;/h1,3-8H2,2H3;3*1H;/q;;;;+3/p-3
SMILES CCCCCCCC[Sn](Cl)(Cl)Cl
IUPAC Name trichloro(octyl)stannane
Molecular Weight (g/mol) 338.29
Formula Weight 337.9418
Percent Purity >95%
Chemical Name or Material n-Octyltin-trichloride
Grade GC