Learn More
Tributyl Trimellitate, TRC
Specifications
Specifications
| Chemical Name or Material | Tri-n-butyl Trimellitate |
| CAS | 1726-23-4 |
| Molecular Formula | C21 H30 O6 |
| Quantity | 500 mg |
| Synonym | 1,2,4-Benzenetricarboxylic acid','tributyl ester (8CI,9CI), Trimellitic acid','tributyl ester (6CI), 1,2,4-Tributyl 1,2,4-benzenetricarboxylate, Monocizer TBTM, Tri-n-butyl trimellitate, Tributyl trimellitate, Trimellitic acid tri-n-butyl ester, Tri-n-butyl Trimellitate |
| InChI Formula | InChI=1S/C21H30O6/c1-4-7-12-25-19(22)16-10-11-17(20(23)26-13-8-5-2)18(15-16)21(24)27-14-9-6-3/h10-11,15H,4-9,12-14H2,1-3H3 |
| SMILES | CCCCOC(=O)c1ccc(C(=O)OCCCC)c(c1)C(=O)OCCCC |
| IUPAC Name | tributyl benzene-1,2,4-tricarboxylate |
| Molecular Weight (g/mol) | 378.4593 |
| Formula Weight | 378.2042 |