Learn More
Tribenzothiophene, TRC
Product Code. 30452961
Change view
Click to view available options
Quantity:
10 mg
100 mg
Unit Size:
10mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30452961 10 mg 10mg
30450150 100 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30452961 Supplier Toronto Research Chemicals Supplier No. TRCT76785010MG
 Restricted Item
Estimated Shipment: 11-11-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 30796-92-0
Molecular Formula C14H8S
Molecular Weight (g/mol) 208.28
Synonym 4,5-Epithiophenanthrene, Benzo[def]dibenzothiophene, Phenanthro[4,5-bcd]thiophene
SMILES c1cc2ccc3cccc4sc(c1)c2c34
Specifications
Quantity 10 mg
InChI Formula InChI=1S/C14H8S/c1-3-9-7-8-10-4-2-6-12-14(10)13(9)11(5-1)15-12/h1-8H
Formula Weight 208.0347
Chemical Name or Material Tribenzothiophene