Learn More
BRAND™ Triangular Wedge-Shape PTFE Magnetic Stirring Bars
Shop All Brand Products
Click to view available options
Length (Metric):
25 mm
35 mm
40 mm
50 mm
55 mm
80 mm
Diameter (Metric):
8 mm
10 mm
12 mm
14 mm
16 mm
Unit Size:
Pack of 10
Description
- Ideal for dissolving solids and mixing of sediments due to scraping action
- Angular surface creates high turbulence even at low speeds
- Provided in packs of 10
Specifications
Specifications
| Description | Triangular wedge-shaped |
| Diameter (Metric) | 16 mm |
| Length (Metric) | 80 mm |
| Material | PTFE |
| Quantity | 10/Pk. |
| Shape | Triangle |