Learn More
Copan Italia™ Cell Spreader
Click to view available options
Quantity:
1000/Pk.
500 Pk.
Material:
Plastic
Polystyrene
Unit Size:
Pack of 1000
Pack of 500
Description
- 171mm
- T shape
- Sterile
- Blue
Specifications
Specifications
| Disposable | Single-use |
| Color | Blue |
| Length (Metric) | 171 mm |
| Quantity | 500 Pk. |
| Shape | T |
| Material | Plastic |
| Product Type | Cell Spreader |
| Sterility | Sterile |