missing translation for 'onlineSavingsMsg'
Learn More

Triallylamine, 97%

Product Code. p-7081900
Change view
Click to view available options
Quantity:
250 g
1000 g
Unit Size:
1000g
250g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11469992 250 g 250g
11449992 1000 g 1000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11469992 Supplier Thermo Scientific Alfa Aesar Supplier No. A12081.30
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 102-70-5 | C9H15N | 137.226 g/mol

It is employed in the production of other chemicals. It is also mainly applied in organic synthesis and resin modifiers. It is used in the cross linking of high absorbent and the intermediates of ion exchange resin. It can also be employed in producing polyester activator and the evocating agent of butadiene polymerization.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed in the production of other chemicals. It is also mainly applied in organic synthesis and resin modifiers. It is used in the cross linking of high absorbent and the intermediates of ion exchange resin. It can also be employed in producing polyester activator and the evocating agent of butadiene polymerization.

Solubility
Not miscible or difficult to mix in water.

Notes
Store away from strong oxidizing agents and acids. Protect against electrostatic charges and keep away from ignition sources.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 102-70-5
Molecular Formula C9H15N
Molecular Weight (g/mol) 137.226
MDL Number MFCD00026093
InChI Key VPYJNCGUESNPMV-UHFFFAOYSA-N
Synonym triallylamine, tris 2-propenyl amine, 2-propen-1-amine, n,n-di-2-propenyl, triallyl amine, unii-b6n19xc04r, ccris 4876, n,n-diallylprop-2-en-1-amine, n,n-di-2-propenyl-2-propen-1-amine, ch2=chch2 3n, 4-04-00-01061 beilstein handbook reference
PubChem CID 7617
IUPAC Name N,N-bis(prop-2-enyl)prop-2-en-1-amine
SMILES C=CCN(CC=C)CC=C

Specifications

Melting Point -70°C
Density 0.79
Boiling Point 150°C to 151°C
Flash Point 31°C (87°F)
Odor Amine-like
Refractive Index 1.451
Quantity 250 g
UN Number UN2610
Solubility Information Not miscible or difficult to mix in water.
Formula Weight 137.23
Percent Purity 98%
Chemical Name or Material Triallylamine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.