Learn More
Tri-n-butyl-d27-tin Chloride, CDN
Click to view available options
Quantity:
0.1 g
0.25 g
Unit Size:
0.1g
0.25g
Specifications
Specifications
| Chemical Name or Material | Tributyltin chloride-d27 |
| CAS | 1257647-76-9 |
| Molecular Formula | C12 2H27 Cl Sn |
| Quantity | 0.1 g |
| Synonym | Tributyltin chloride (D27), Tributyltin Chloride D27 |
| InChI Formula | InChI=1 S/3C4H9.ClH.Sn/c3*1-3-4-2;;/h3*1,3-4H2,2H3;1 H;/q;;;;+1/p-1/i3*1D2,2D3,3D2,4D2;; |
| SMILES | [2 H]C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])[Sn](Cl)(C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])[2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])[2 H] |
| IUPAC Name | chloro-tris(1,1,2,2,3,3,4,4,4-nonadeuteriobutyl)stannane |
| Molecular Weight (g/mol) | 352.67 |
| Formula Weight | 353.2518 g/mol |
| Show More |