Learn More
Tri(m-tolyl)phosphine, 98+%
Product Code. 11438316
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
Product Code. 11438316
Brand: Thermo Scientific Alfa Aesar A15372.03
Request Bulk or Custom Format
CAS: 6224-63-1 | C21H21P | 304.373 g/mol
Tri(m-tolyl)phosphine is used in suzuki reaction. It acts as a ligand which is used in organometallic chemistry.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Tri(m-tolyl)phosphine is used in suzuki reaction. It acts as a ligand which is used in organometallic chemistry.
Solubility
Insoluble in water.
Notes
Air Sensitive. Store away from incompatible materials such as oxidizing agents.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 6224-63-1
Molecular Formula C21H21P
Molecular Weight (g/mol) 304.373
MDL Number MFCD00008532
InChI Key LFNXCUNDYSYVJY-UHFFFAOYSA-N
Synonym tri-m-tolylphosphine, tris 3-methylphenyl phosphine, tri m-tolyl phosphine, phosphine, tris 3-methylphenyl, tris m-tolyl phosphine, phosphine, tri-m-tolyl, tris 3-methylphenyl phosphane, tris 3-tolyl phosphine, tri-m-tolyl-phosphane, tri-m-tolylphosphane
PubChem CID 80362
IUPAC Name tris(3-methylphenyl)phosphane
SMILES CC1=CC(=CC=C1)P(C2=CC=CC(=C2)C)C3=CC=CC(=C3)C
Specifications
Melting Point 99°C to 101°C
Quantity 1 g
Beilstein 658863
Sensitivity Air Sensitive
Solubility Information Insoluble in water.
Formula Weight 304.37
Percent Purity ≥98%
Chemical Name or Material Tri(m-tolyl)phosphine