missing translation for 'onlineSavingsMsg'
Learn More

Tri(m-tolyl)phosphine, 98+%

Product Code. 11448316
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 11448316

Brand: Thermo Scientific Alfa Aesar A15372.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 6224-63-1 | C21H21P | 304.373 g/mol

Tri(m-tolyl)phosphine is used in suzuki reaction. It acts as a ligand which is used in organometallic chemistry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tri(m-tolyl)phosphine is used in suzuki reaction. It acts as a ligand which is used in organometallic chemistry.

Solubility
Insoluble in water.

Notes
Air Sensitive. Store away from incompatible materials such as oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6224-63-1
Molecular Formula C21H21P
Molecular Weight (g/mol) 304.373
MDL Number MFCD00008532
InChI Key LFNXCUNDYSYVJY-UHFFFAOYSA-N
Synonym tri-m-tolylphosphine, tris 3-methylphenyl phosphine, tri m-tolyl phosphine, phosphine, tris 3-methylphenyl, tris m-tolyl phosphine, phosphine, tri-m-tolyl, tris 3-methylphenyl phosphane, tris 3-tolyl phosphine, tri-m-tolyl-phosphane, tri-m-tolylphosphane
PubChem CID 80362
IUPAC Name tris(3-methylphenyl)phosphane
SMILES CC1=CC(=CC=C1)P(C2=CC=CC(=C2)C)C3=CC=CC(=C3)C

Specifications

Melting Point 99°C to 101°C
Quantity 5 g
Beilstein 658863
Sensitivity Air Sensitive
Solubility Information Insoluble in water.
Formula Weight 304.37
Percent Purity ≥98%
Chemical Name or Material Tri(m-tolyl)phosphine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.