missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Treprostinil
GREENER_CHOICE

Product Code. 18714069
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
This item is not returnable. View return policy

Product Code. 18714069

Brand: Tocris Bioscience™ 5349/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potent prostacyclin (PGI2) analog

Treprostinil is a potent prostacyclin (PGI2) analog (EC50 values are 0.6, 1.9 and 6.2 nM at DP1, IP and EP2 receptors respectively). Causes vasodilation of human pulmonary arteries and inhibits NFκB nuclear translocation in human alveolar macrophages in vitro.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-[[(1R,2R,3aS,9aS)-2,3,3a,4,9,9a-Hexahydro-2-hydroxy-1-[(3S)-3-hydroxyoctyl]-1H-benz[f]inden-5-yl]oxy]acetic acid
CAS 81846-19-7
Quantity 10 mg
Target Prostanoid Receptor Agonists
Molecular Formula C23H34O5
Formula Weight 390.51
Purity >98%
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.