Learn More
Tocris Bioscience™ Treprostinil
GREENER_CHOICE
Potent prostacyclin (PGI2) analog
Brand:  Tocris Bioscience™ 5349/10
Encompass
Product Code. 15746685
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products »
Description
Description
Treprostinil is a potent prostacyclin (PGI2) analog (EC50 values are 0.6, 1.9 and 6.2 nM at DP1, IP and EP2 receptors respectively). Causes vasodilation of human pulmonary arteries and inhibits NFκB nuclear translocation in human alveolar macrophages in vitro.
Specifications
Specifications
2-[[(1R,2R,3aS,9aS)-2,3,3a,4,9,9a-Hexahydro-2-hydroxy-1-[(3S)-3-hydroxyoctyl]-1H-benz[f]inden-5-yl]oxy]acetic acid
81846-19-7
10 mg