missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Trazodone-d6 HCl (propyl-d6), CDN
Click to view available options
Quantity:
0.005 g
0.01 g
Unit Size:
0.01g
Specifications
Specifications
| Chemical Name or Material | Trazodone-D6 Hydrochloride |
| CAS | 1181578-71-1 |
| Molecular Formula | C19 2H6 H16 Cl N5 O . Cl H |
| Quantity | 0.01 g |
| Synonym | Trazodone-D6 Hydrochloride |
| InChI Formula | InChI=1 S/C19H22ClN5O.ClH/c20-16-5-3-6-17(15-16)23-13-11-22(12-14-23)8-4-10-25-19(26)24-9-2-1-7-18(24)21-25;/h1-3,5-7,9,15 H,4,8,10-14H2;1 H/i4D2,8D2,10D2; |
| SMILES | Cl.[2 H]C([2 H])(N1CCN(CC1)c2cccc(Cl)c2)C([2 H])([2 H])C([2 H])([2 H])N3N=C4C=CC=CN4C3=O |
| IUPAC Name | 2-[3-[4-(3-chlorophenyl)piperazin-1-yl]-1,1,2,2,3,3-hexadeuteriopropyl]-[1,2,4 ]triazolo[4,3-a]pyridin-3-one;hydrochloride |
| Molecular Weight (g/mol) | 414.36 |
| Formula Weight | 413.1656 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction