Learn More
JULABO™ Standard Carrier Tray for Erlenmeyer Flasks
Product Code. 10319530
Click to view available options
Holds:
11 Erlenmeyer Flasks, 250 to 300 mL
15 Erlenmeyer Flasks, 200 mL
32 Erlenmeyer Flasks, 50 mL
45 Erlenmeyer Flasks, 25 mL
5 Erlenmeyer Flasks, 1000 mL
8 Erlenmeyer Flasks, 500 mL
Unit Size:
Each
Product Code. 10319530
Brand: JULABO™ 8970363
Only null left
Add to basket
Add to basket
For use with Erlenmeyer flasks. Suitable for JULABO™ SW22 / SW23.
  • Metal Construction
  • For use with JULABO™ SW22/SW23
  • Several sizes available
  • Choose a model to hold as few as 5 x 1000 mL Erlenmeyer Flasks or as many as 45 x 25 mL
Specifications
For Use With (Equipment) SW22, SW23 shaking water bath
Type Carrier Tray
Holds 15 Erlenmeyer Flasks, 200 mL