Learn More
Stevanato Group Tray Rotation Tool from DWK Life Sciences™
Product Code. 17870535
Change view
Click to view available options
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Unit Size
17870535 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17870535 Supplier Stevanato Group Supplier No. SP47004493
  • ONLINE EXCLUSIVE PRICE
    467.00€
    415.67€ / Pack of 1
    Save 51.33€
    10% Off
Estimated Shipment: 10-12-2026
Tray rotation tool for use with EZ-fill™ Vials.
  • Tray rotation tool for vial reorientation (neck down to neck up)
  • Improves workflow by assisting vial staging
Specifications
Product Type Tray Rotation Tool