Learn More
Bochem™ Transport Container Seal
Product Code. 10411123
Click to view available options
For Use With (Equipment):
1L Transport Container
Unit Size:
Each
You may only order up to the available stock. For more details call customer services on +351 21 425 33 50.
Product Code. 10411123
Brand: Bochem™ 18300SS
Only null left
Add to basket
Add to basket
You may only order up to the available stock. For more details call customer services on +351 21 425 33 50.
Bochem™ Transport Container Seal is a silicone seal for use with a safety transport container.
  • Silicone
  • For use with transport container
Specifications
For Use With (Equipment) 1L Transport Container
Diameter (Metric) Outside 130 mm
Material Silicone