Learn More
Amsta Rostfritt Stainless Steel Transport Bottle Churn With Lid
Click to view available options
Capacity (Metric):
25 L
30 L
40 L
50 L
Unit Size:
Each
Description
- Stainless steel 304 /1.4301
Specifications
Specifications
| Product Type | Pail |
| Capacity (Metric) | 50 L |
| Color | Silver |
| Material | Stainless Steel |