Learn More
Fisherbrand™ Non-sterile Transfer Pipettes, Transparent LDPE
Product Code. 13499108 Shop All Fisherbrand Products
Change view
Click to view available options
Volume (Metric):
1 mL
3 mL
4 mL
7 mL
10 mL
Drops/mL:
18
20
25
30
50
Graduated:
No
Yes
Tip Style:
Extended
Extra Long
Jumbo
Standard
Thin Stem
Unit Size:
Pack of 100
Pack of 200
Pack of 400
Pack of 500
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Drops/mL Graduated Tip Style unitSize
13499108 1 mL 30 Yes Standard Pack of 500
13469118 1 mL 30 No Standard Pack of 400
13439118 1 mL 30 Yes Extended Pack of 500
13439108 3 mL 25 Yes Standard Pack of 500
13459118 4 mL 30 No Thin Stem Pack of 500
13459108 4 mL 50 No Standard Pack of 500
13449108 7 mL 20 No Extra Long Pack of 100
13449118 10 mL 18 No Jumbo Pack of 200
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Product Code. 13499108
Only null left
Add to basket
Edge
Add to basket
Prevent cross contamination. Fisherbrand™ Non-sterile Transfer Pipets, Transparent LDPE are made of shatterproof, flexible plastic and are designed to replace glass Pasteur pipets. Graduated and non-graduated pipets available in a range of sizes and styles, including jumbo, thin stem and extra long.
  • Built-in bulb eliminates the need for plugging, packaging and autoclaving
  • Flexible stem bends to sample from narrow or small tubes
  • Low affinity surface reduces loss of cells and valuable proteins due to binding
  • Soft plastic pipets made of non-toxic low-density polyethylene (LDPE)
  • Non-sterile, transparent and fully recyclable
Specifications
Material LDPE
Drops/mL 30
Graduated Yes
Graduations 0.25 mL
Quantity 500 Pack
Tip Style Standard
Volume (Metric) Dispense 33 μL drop
Volume (Metric) 1 mL
Length (Metric) 150 mm
Type Transfer Pipet