Learn More
Fisherbrand™ Translucent Silicone Tubing
Use this tubing with peristaltic pumps. Fisherbrand™ Translucent Silicone Tubing is ideal for pharmaceutical, biotechnology and sterile fluid transfer applications.
Brand: Fisherbrand™
Additional Details : Weight : 2.97000kg
Description
- Translucent silicone tubing
- FDA compliant (food)
Specifications
FDA, BFR and WRAS approved | |
Silicone | |
25 m | |
Peristaltic pumps | |
-40°C | |
Translucent | |
250°C |
8 mm | |
14 mm | |
3 mm | |
Pharmaceutical , biotechnology, transfer sterile fluids | |
Sterile | |
Flexible |