Learn More
Fisherbrand™ Translucent Silicone Tubing
Product Code. 11733416 Shop All Fisherbrand Products
Click to view available options
Diameter (Metric) Inner:
1 mm
2 mm
3 mm
4 mm
5 mm
6 mm
7 mm
8 mm
10 mm
Diameter (Metric) Outer:
3 mm
6 mm
7 mm
8 mm
9 mm
10 mm
12 mm
14 mm
15 mm
Overall Length (Metric):
25 m
5 m
Unit Size:
25m
5m
Each
Product Code. 11733416
Edge
Use this tubing with peristaltic pumps. Fisherbrand™ Translucent Silicone Tubing is ideal for pharmaceutical, biotechnology and sterile fluid transfer applications.
  • Translucent silicone tubing
  • FDA compliant (food)
Specifications
Certifications/Compliance FDA, BFR and WRAS approved
Diameter (Metric) Inner 6 mm
Material Silicone
Diameter (Metric) Outer 12 mm
Overall Length (Metric) 25 m
Thickness (Metric) 3 mm
For Use With Peristaltic pumps
For Use With (Application) Pharmaceutical , biotechnology, transfer sterile fluids
Min. Operating Temperature (Metric) -40°C
Sterility Sterile
Color Translucent
Flexibility Flexible
Max. Operating Temperature (Metric) 250°C