Learn More
Translucent Crucibles
Product Code. 12807733
Click to view available options
Diameter (Metric) Outer:
29mm
38mm
43mm
57mm
67mm
82mm
Unit Size:
Each
Product Code. 12807733
Brand: Yorlab 10444/20
Only null left
Specifications
Color Translucent
Material Silica
Capacity (Metric) 80 mL
Diameter (Metric) Outer 67mm
Height (Metric) 45mm
Product Type Crucible