missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ TranslationBlocker Rat High Affinity Nerve Growth Factor Receptor (TrkA) siRNA

Product Code. 16079211
Change view
Click to view available options
Quantity:
10 nmol
2 nmol
Unit Size:
10nmol
2nmol
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16079211 2 nmol 2nmol
16069211 10 nmol 10nmol
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16079211 Supplier enQuireBio™ Supplier No. QX342nmol

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Blocks transcription and translation of High Affinity Nerve Growth Factor Receptor (TrkA) from Rat.

Specifications

Accession Number NP_067600
Cell Lines Tested Pc12 Rat Pheocromocytoma Cells from the Adrenal Medulla and Rat Primary Neurons.
Content And Storage Store Lyophilized siRNA duplex at 4°C. Reconsituted product may be stored at -20°C for up to 1 month.
Gene Symbol Ntrk1
Gene ID (Entrez) 59109
Product Type siRNA
Quantity 2 nmol
Synonym TrkA, High Affinity NGFR, NGFR, Trk
Target Rat
Form Lyophilized
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.