missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ TranslationBlocker Rat Estrogen receptor beta siRNA

Product Code. 16099171
Change view
Click to view available options
Quantity:
10 nmol
2 nmol
Unit Size:
10nmol
2nmol
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16099171 2 nmol 2nmol
16089171 10 nmol 10nmol
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16099171 Supplier enQuireBio™ Supplier No. QX172nmol

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Blocks transcription and translation of Estrogen receptor beta from Rat.

Specifications

Accession Number NP_036886
Cell Lines Tested Rat Primary Osteoblasts.
Content And Storage Store Lyophilized siRNA duplex at 4°C. Reconsituted product may be stored at -20°C for up to 1 month.
Gene Symbol Esr2
Gene ID (Entrez) 25149
Product Type siRNA
Quantity 2 nmol
Synonym ERbeta, Erb2, ER-beta
Target Rat
Form Lyophilized
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.