Learn More
enQuireBio™ TranslationBlocker Human PD-L2 / CD273 siRNA
Product Code. 16019261
Change view
Click to view available options
Quantity:
10 nmol
2 nmol
Unit Size:
10nmol
2nmol
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16019261 2 nmol 2nmol
16009261 10 nmol 10nmol
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 16019261 Supplier enQuireBio™ Supplier No. QX82nmol
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Blocks transcription and translation of PD-L2 / CD273 from Human.
Specifications
Accession Number NP_079515
Cell Lines Tested Hep3b Human Hepatocellular Carinoma Cells, RKO Human Colon Cancer Cells, and Primary Dendritic Cells (DCs) isolated from PBMCs.
Content And Storage Store Lyophilized siRNA duplex at 4°C. Reconsituted product may be stored at -20°C for up to 1 month.
Gene Symbol PDCD1LG2
Gene ID (Entrez) 80380
Product Type siRNA
Quantity 2 nmol
Synonym PDL2, CD273, Programmed Cell Death 1 Ligand 2, PDCD1L2, B7-DC, B7DC, PD-1 L2, PD-1 Ligand 2, PDCD1 Ligand 2
Target Human
Form Lyophilized