Learn More
enQuireBio™ TranslationBlocker Human MyD88 siRNA
Product Code. 16079191
Change view
Click to view available options
Quantity:
10 nmol
2 nmol
Unit Size:
10nmol
2nmol
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16079191 2 nmol 2nmol
16069191 10 nmol 10nmol
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 16079191 Supplier enQuireBio™ Supplier No. QX252nmol
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Blocks transcription and translation of MyD88 from Human.
Specifications
Accession Number NP_002459
Cell Lines Tested 293T Human Embryonic Kidney Cells with Large T Antigen.
Content And Storage Store Lyophilized siRNA duplex at 4°C. Reconsituted product may be stored at -20°C for up to 1 month.
Gene Symbol MYD88
Gene ID (Entrez) 4615
Product Type siRNA
Quantity 2 nmol
Synonym MYD88D, Myeloid differentiation primary response protein, Toll-Receptor Signaling Cascade
Target Human
Form Lyophilized