missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ TranslationBlocker Human FADD siRNA

Product Code. 16099231
Change view
Click to view available options
Quantity:
10 nmol
2 nmol
Unit Size:
10nmol
2nmol
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16099231 2 nmol 2nmol
16089231 10 nmol 10nmol
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16099231 Supplier enQuireBio™ Supplier No. QX442nmol

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products

This item is not returnable. View return policy

Blocks transcription and translation of FADD from Human.

Specifications

Accession Number NP_003815
Cell Lines Tested 293 Human Embryonic Kidney Cells with NF-kappa B and Flag tagged DR4 constructs.
Content And Storage Store Lyophilized siRNA duplex at 4°C. Reconsituted product may be stored at -20°C for up to 1 month.
Gene Symbol FADD
Gene ID (Entrez) 8772
Product Type siRNA
Quantity 2 nmol
Synonym Fas Associated Via Death Domain, Fas-Associating Death Domain-Containing Protein, Growth-Inhibiting Gene 3 Protein, Fas (TNFRSF6)-Associated via Death Domain, Protein FADD, MORT1, FAS-Associated Death Domain Protein
Target Human
Form Lyophilized
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.