missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ TranslationBlocker Human Estrogen receptor siRNA

Product Code. 16029171
Click to view available options
Quantity:
10 nmol
2 nmol
Unit Size:
10nmol
2nmol
This item is not returnable. View return policy

Product Code. 16029171

Brand: enQuireBio™ QX1410nmol

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products

This item is not returnable. View return policy

Blocks transcription and translation of Estrogen receptor from Human.

Specifications

Accession Number NP_000116
Cell Lines Tested MCF7 Human Adenocarcinoma Cells from Breast Cancer.
Content And Storage Store Lyophilized siRNA duplex at 4°C. Reconsituted product may be stored at -20°C for up to 1 month.
Gene Symbol ESR1
Gene ID (Entrez) 2099
Product Type siRNA
Quantity 10 nmol
Synonym ESRA, ESR, NR3A1, ER, ESTRR, Era
Target Human
Form Lyophilized
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.