Learn More
enQuireBio™ TranslationBlocker Human Caspase-1 siRNA
Product Code. 16049221
Change view
Click to view available options
Quantity:
1 nmol
5 nmol
Unit Size:
1nmol
5nmol
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16049221 1 nmol 1nmol
16059221 5 nmol 5nmol
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 16049221 Supplier enQuireBio™ Supplier No. QX381nmol
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Blocks transcription and translation of Caspase-1 from Human.
Specifications
Accession Number NP_001214
Cell Lines Tested Human Primary Keratinocytes.
Content And Storage Store Lyophilized siRNA duplex at 4°C. Reconsituted product may be stored at -20°C for up to 1 month.
Gene Symbol CASP1
Gene ID (Entrez) 834
Product Type siRNA
Quantity 1 nmol
Synonym Interleukin 1, Beta, Convertase, ICE, Caspase 1
Target Human
Form Lyophilized