missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Novus Biologicals™ Transferrin Native Protein
Shop alle Bio Techne produkter
Klik for at se tilgængelige muligheder
Quantity:
10 mg
Pakningsstørrelse:
10mg
Tekniske data
Tekniske data
| Accession Number | Q06AH7 |
| For Use With (Application) | Western Blot, ELISA, SDS-PAGE |
| Formulation | Lyophilized from 0.2 M ammonium bicarbonate. |
| Gene ID (Entrez) | 7018 |
| Quantity | 10 mg |
| Storage Requirements | Store at 4°C. |
| Gene Alias | Beta-1 metal-binding globulin, DKFZp781D0156, EC 3.4.21, PRO1557, PRO2086, serotransferrin, Siderophilin, transferrin |
| Gene Symbol | TF |
| Reconstitution | Reconstitute with 1.0 mL deionized water. |
| Inhibitor Properties | This product was lyophilized from 0.2 M ammonium bicarbonate for use in cell culture and assays. Transferrin has been tested in SDS-Page and is also suitable for use as antigen or ligand in immunochemical reactions, as a control or standard in assays, for conjugation and most other immunological methods requiring highly purified proteins. |
| Vis mere |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion