missing translation for 'onlineSavingsMsg'
Learn More
Learn More
trans-Stilbene-d12, CDN
Click to view available options
Quantity:
0.25 g
0.5 g
Unit Size:
0.25g
0.5g
Specifications
Specifications
| Chemical Name or Material | trans-Stilbene-d12 |
| CAS | 16341-52-9 |
| Molecular Formula | C14 2H12 |
| Quantity | 0.25 g |
| Synonym | Benzene-1,2,3,4,5-d5, 6,6'-[(1 E)-1,2-ethenediyl-1,2-d2]bis-, Benzene-d5, 6,6'-(1,2-ethenediyl-1,2-d2)bis-, (E)-, Benzene-d5, 6,6'-[(1 E)-1,2-ethenediyl-1,2-d2]bis- (9 CI), Stilbene-d12, (E)- (8 CI), 6,6'-[(1 E)-1,2-Ethenediyl-1,2-d2]bis[benzene-1,2,3,4,5-d5], trans-Stilbene-d12 |
| InChI Formula | InChI=1 S/C14H12/c1-3-7-13(8-4-1)11-12-14-9-5-2-6-10-14/h1-12 H/b12-11+/i1D,2 D,3 D,4 D,5 D,6 D,7 D,8 D,9 D,10 D,11 D,12 D |
| SMILES | [2 H]\C(=C(\[2 H])/c1c([2 H])c([2 H])c([2 H])c([2 H])c1[2 H])\c2c([2 H])c([2 H])c([2 H])c([2 H])c2[2 H] |
| IUPAC Name | 1,2,3,4,5-pentadeuterio-6-[(E)-1,2-dideuterio-2-(2,3,4,5,6-pentadeuteriophenyl)ethenyl]benzene |
| Molecular Weight (g/mol) | 192.32 |
| Formula Weight | 192.1692 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction