missing translation for 'onlineSavingsMsg'
Learn More

trans-Dihydro Lisuride, TRC

Product Code. 30387495
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30387495 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30387495 Supplier Toronto Research Chemicals Supplier No. TRCD44949510MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Terguride
CAS 37686-84-3
Molecular Formula C20 H28 N4 O
Quantity 10 mg
Synonym Urea, N,N-diethyl-N'-[(8α)-6-methylergolin-8-yl]-, Ergoline, urea deriv., Indolo[4,3-fg]quinoline, urea deriv., N,N-Diethyl-N'-[(8α)-6-methylergolin-8-yl]urea, (+)-(5R,8S,10R)-Terguride, (+)-Terguride, (5R,8S,10R)-Terguride, 6-Methyl-8α-(diethylcarbamoylamino)ergoline, 9,10α-Dihydrolisuride, N,N-Diethyl-N'-(D-6-methyl-10α-isoergolin-8-yl)urea, TDHL, Tergurid, Terguride, trans-Dihydrolisuride
InChI Formula InChI=1S/C20H28N4O/c1-4-24(5-2)20(25)22-14-10-16-15-7-6-8-17-19(15)13(11-21-17)9-18(16)23(3)12-14/h6-8,11,14,16,18,21H,4-5,9-10,12H2,1-3H3,(H,22,25)/t14-,16+,18+/m0/s1
SMILES CCN(CC)C(=O)N[C@H]1C[C@H]2[C@@H](Cc3c[nH]c4cccc2c34)N(C)C1
IUPAC Name 3-[(6aR,9S,10aR)-7-methyl-6,6a,8,9,10,10a-hexahydro-4H-indolo[4,3-fg]quinoline-9-yl]-1,1-diethylurea
Molecular Weight (g/mol) 340.46
Formula Weight 340.2263
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.