Learn More
trans-Cotinine Amide, TRC
Product Code. 30364142
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30364142 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30364142 Supplier Toronto Research Chemicals Supplier No. TRCC72510025MG
 Restricted Item
Estimated Shipment: 17-08-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material trans-Cotinine Amide
CAS 939437-34-0
Molecular Formula C11H13N3O2
Quantity 25 mg
InChI Formula InChI=1S/C11H13N3O2/c1-14-9(15)5-8(11(12)16)10(14)7-3-2-4-13-6-7/h2-4,6,8,10H,5H2,1H3,(H2,12,16)/t8-,10+/m0/s1
SMILES CN1[C@@H]([C@H](CC1=O)C(=O)N)c2cccnc2
IUPAC Name (2S,3S)-1-methyl-5-oxo-2-pyridin-3-ylpyrrolidine-3-carboxamide
Molecular Weight (g/mol) 219.24
Formula Weight 219.1008