Learn More
trans-5-Decen-1-ol, 96%
Product Code. 11312617
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Product Code. 11312617
Brand: Thermo Scientific Alfa Aesar L11768.06
Request Bulk or Custom Format
CAS: 56578-18-8 | C10H20O | 156.27 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 56578-18-8
Molecular Formula C10H20O
Molecular Weight (g/mol) 156.27
MDL Number MFCD00056187
InChI Key WYPQHXVMNVEVEB-AATRIKPKSA-N
Synonym trans-5-decen-1-ol, e-5-decen-1-ol, trans-5-decenol, 5-decen-1-ol, 5e, 5-decen-1-ol, e, decan-5-en-1-ol, e-5-decenol, unii-m709v74ozv, 5-decen-1-ol, epa pesticide chemical code 078038
PubChem CID 5283292
IUPAC Name (E)-dec-5-en-1-ol
SMILES CCCC\C=C\CCCCO
Specifications
Boiling Point 113°C to 118°C (11 mmHg)
Flash Point 62°C (143°F)
Refractive Index 1.452
Quantity 5 g
Beilstein 1840630
Formula Weight 156.27
Percent Purity 96%
Chemical Name or Material trans-5-Decen-1-ol