Learn More
trans-4-Aminocyclohexanecarboxylic acid, 97%
Product Code. 11481720
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Produktkod. 11481720
missing translation for 'mfr': Thermo Scientific Alfa Aesar H27489.06
Request Bulk or Custom Format
Denna artikel kan inte returneras. Se returpolicy
CAS: 3685-25-4 | C7H13NO2 | 143.186 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 3685-25-4
Molecular Formula C7H13NO2
Molecular Weight (g/mol) 143.186
MDL Number MFCD00209953
InChI Key DRNGLYHKYPNTEA-UHFFFAOYSA-N
Synonym 4-aminocyclohexanecarboxylic acid, trans-4-aminocyclohexanecarboxylic acid, cis-4-aminocyclohexanecarboxylic acid, cis-4-aminocyclohexane carboxylic acid, cis-4-amino-1-cyclohexanecarboxylic acid, trans-4-amino-cyclohexanecarboxylic acid, cis-4-aminocyclohexanecarboxylicacid, cyclohexanecarboxylic acid, 4-amino-, trans, trans-4-aminocyclohexane carboxylic acid, 1s,4s-4-aminocyclohexanecarboxylic acid
PubChem CID 171560
IUPAC Name 4-aminocyclohexane-1-carboxylic acid
SMILES C1CC(CCC1C(=O)O)N
Specifications
Melting Point >300°C
Quantity 5 g
Formula Weight 143.19
Percent Purity 97%
Chemical Name or Material trans-4-Aminocyclohexanecarboxylic acid