Learn More
trans-2-[trans-2-(1,3,3-Trimethyl-2-indolinylidene)ethylidene]-1-indanone, 96%, Thermo Scientific™
Product Code. 15465135
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
Product Code. 15465135
Brand: Thermo Scientific Alfa Aesar H31932.MD
Request Bulk or Custom Format
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 53704-23-7
Molecular Formula C22H21NO
Molecular Weight (g/mol) 315.416
MDL Number MFCD06409584
InChI Key RMZNRQVMHQVFHH-UHFFFAOYSA-N
Synonym 2e-2-2-2e-1,3,3-trimethylindol-2-ylidene ethylidene-3h-inden-1-one, 2e-2-2e-2-1,3,3-trimethyl-1,3-dihydro-2h-indol-2-ylidene ethylidene indan-1-one, e-2-2-e-1,3,3-trimethylindolin-2-ylidene ethylidene-2,3-dihydro-1h-inden-1-one, trans-2-trans-2-1,3,3-trimethyl-2-indolinylidene ethylidene-1-indanone, e-2-e-2-1,3,3-trimethylindolin-2-ylidene ethylidene-2,3-dihydro-1h-inden-1-one, 2-2-1,3,3-trimethylindolin-2-ylidene ethylidene-2,3-dihydro-1h-inden-1-one
PubChem CID 53426998
IUPAC Name 2-[2-(1,3,3-trimethylindol-2-ylidene)ethylidene]-3H-inden-1-one
SMILES CC1(C2=CC=CC=C2N(C1=CC=C3CC4=CC=CC=C4C3=O)C)C
Specifications
Melting Point 184°C to 187°C
Quantity 250 mg
Formula Weight 315.42
Percent Purity 96%
Chemical Name or Material trans-2-[trans-2-(1,3,3-Trimethyl-2-indolinylidene)ethylidene]-1-indanone