Learn More
trans-2,3,4-Trimethoxycinnamic acid, 99%, Thermo Scientific™
Product Code. 11321387
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11321387 5 g 5g
11331387 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11321387 Supplier Thermo Scientific Alfa Aesar Supplier No. L01703.06
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 33130-03-9
Molecular Formula C12H14O5
Molecular Weight (g/mol) 238.239
MDL Number MFCD00014376
InChI Key ZYOPDNLIHHFGEC-FNORWQNLSA-N
Synonym 2,3,4-trimethoxycinnamic acid, trans-2,3,4-trimethoxycinnamic acid, 3-2,3,4-trimethoxyphenyl acrylic acid, 2e-3-2,3,4-trimethoxyphenyl prop-2-enoic acid, 2-propenoic acid, 3-2,3,4-trimethoxyphenyl, 3-2,3,4-trimethoxyphenyl propenoic acid, trans-2, 3, 4-trimethoxycinnamate, 2e-3-2,3,4-trimethoxyphenyl-2-propenoic acid, e-3-2,3,4-trimethoxyphenyl acrylic acid, 2e-3-2,3,4-trimethoxyphenyl acrylic acid
PubChem CID 735841
IUPAC Name (E)-3-(2,3,4-trimethoxyphenyl)prop-2-enoic acid
SMILES COC1=C(C(=C(C=C1)C=CC(=O)O)OC)OC
Specifications
Melting Point 172°C to 176°C
Quantity 5 g
Beilstein 2657904
Formula Weight 238.24
Percent Purity 99%
Chemical Name or Material trans-2,3,4-Trimethoxycinnamic acid